Order Entry
Finland
ContactUsLinkComponent
 
Articulated magnifier
  MOIN1611VE
 :  MOINEAU
undefined
Articulated magnifier
  MOIN1611VE
 :  MOINEAU

 

  • Ø:
    50 mm
  • Magnification:
    2,5×
  • Description:
    Third-hand magnifier (500 g)
  • Pk:
    1

 

 

"Third hand" magnifier in mineral glass, on cast iron foot.

Two crocodile clips for the observation of an object without having to hold in hands.