Biotinylated heparin, porcine intestinal mucosa, Calbiochem®
Catalog # 375054-10
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:10 mg
- Description:Heparin, porcine intestinal mucosa, biotin conjugate
Specifications
About this item
Biotinylated heparin contains 5 − 8 mole-percent biotin. Biotin attachment is through a 12-atom spacer arm containing a cleavable disulfide group.