J.T.Baker®, Formalin Neutraliser
Catalog # 3934.5000
Supplier: Avantor
CAS Number:
Specifications
- Pk:5 kg
- Description:Formalin neutraliser
Specifications
About this item
Formalin (dissolved formaldehyde) is used in histopathology laboratories.
- Histology grade powdered chemical
- Contains sodium disulphite min. 80 %
- Stable for two years when bottles are closed and stored at room temperature
Packaging: In a PP drum.