Cut resistant gloves, Showa 541
Supplier: Showa
Certificates
About this item
These seamless knitted HPPE liners with PU coating on palm and fingers provide a high degree of comfort when performing delicate tasks in greasy work environments with a high risk of cuts.
- HPPE lining provides extremely comfortable fit
- Knitwrist secures glove above the wrist for comfort and prevents debris from entering the glove
- Flat-dipped, grey colored PU coating
- Launderable
Typical applications include: Transport, metallurgy, logistics, steel erecting, construction, automotive.
Certifications: EN 388:2016 (4342B); Cat. III
Specifications
- Catalog No:
- 112-5865
- 112-5863
- 112-5864
- 112-5861
- 112-5862
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- ¾ PU coating
- ¾ PU coating
- ¾ PU coating
- ¾ PU coating
- ¾ PU coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)