Order Entry
Finland
ContactUsLinkComponent
Plate sealing film, AeraSeal
Plate sealing film, AeraSeal
Catalog # AC1201-02
Supplier:  OMEGA BIO-TEK
Plate sealing film, AeraSeal
Catalog # AC1201-02
Supplier:  OMEGA BIO-TEK
Specifications
  • Sterile:
    +
  • Description:
    AeraSeal film sheets
  • Thickness:
    114 µm
  • Pk:
    50
Specifications
About this item
AeraSeal is a 114 µm thick, hydrophobic porous film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth.
  • Minimise cross-contamination, spillage and evaporation
  • Allow uniform air and CO2 exchange for all wells, unlike plate lids which favour exchange for wells near the plate edge
Dimensions: 82,6×142,9 for standard size tissue culture plates.