Order Entry
Finland
ContactUsLinkComponent
Silvers skin graft blades
Catalog # SWAN9942
Supplier:  SWANN MORTON
undefined
Silvers skin graft blades
Catalog # SWAN9942
Supplier:  SWANN MORTON
Specifications
  • Description:
    Silvers skin graft blade, stainless steel, sterile
  • Pk:
    10
Specifications
About this item
This custom designed surgical stainless steel Silver's blade has a 11/2 inch (4 cm) postage stamp size cutting edge to be used with the Silvers miniature Skin Graft Knife handles.
  • Double edged
  • Sterile, individually V.P.I. protected in metal foil packets
  • Can also be used for the debridement of wounds
Ideal for harvesting small intricate grafts from areas which would not be accessible with the larger standard knives such as the ear, dorsa of the hands or scalp.
Packaging: Blades are sealed in individual foil packs.