Order Entry
Finland
ContactUsLinkComponent
Anti-IL-16 Polyclonal Antibody
Anti-IL-16 Polyclonal Antibody
  BSBTPB9238
undefined
Anti-IL-16 Polyclonal Antibody
  BSBTPB9238

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IL-16
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Polyclonal antibody for IL 16/IL16 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. IL 16/IL16 information: Molecular Weight: 141752 MW; Subcellular Localization: Interleukin-16: Secreted; Tissue Specificity: Isoform 3 is expressed in hemopoietic tissues, such as resting T-cells, but is undetectable during active T-cell proliferation.

Type: Primary
Antigen: IL-16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human