Anti-HRG/Hprg Polyclonal Antibody
Catalog # BSBTPB9220
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:HRG/Hprg
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for HPRG/HRG detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. HPRG/HRG information: Molecular Weight: 59578 MW; Subcellular Localization: Secreted ; Tissue Specificity: Expressed in macrophages and in malignant cells. Expressed by the liver and secreted in plasma (at protein level).
Type: Primary
Antigen: HRG/Hprg
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human