Order Entry
Finland
ContactUsLinkComponent
Anti-Cyclin B1 Polyclonal Antibody
Anti-Cyclin B1 Polyclonal Antibody
Catalog # BSBTPB9104
Supplier:  Boster Bio
undefined
Anti-Cyclin B1 Polyclonal Antibody
Catalog # BSBTPB9104
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cyclin B1
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for CCNB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CCNB information: Molecular Weight: 48337 MW; Subcellular Localization: Cytoplasm. Nucleus. Cytoplasm, cytoskeleton, microtubule organizing center, centrosome.

Type: Primary
Antigen: Cyclin B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human