Amikacin disulphate
Catalog # ACRO455190050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤13 % (110°C, 3 h, 5 mm Hg)
- pH:2 to 4 (1 % in water)
- Titration with NaOH:≥87 %
Specification Test Results
About this item
:
:781.77 g/mol
Melting Pt:220…230 °C
Storage Temperature:Refrigerator
CAS Number:39831-55-5
Formula:C₂₂H₄₃N₅O₁₃·2H₂SO₄
MDL Number:MFCD00167475