
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:330645-87-9
:MFCD04973268
Formula:C₁₁H₁₅ClF₆N₅OP
MDL Number:MFCD12031645
:413.69 g/mol
Melting Pt:185…190 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:185 to 191 °C
- Loss on drying:≤0.5 %
- HPLC:≥97.5 %