2-Octyne 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2809-67-8
:
Formula:C₈H₁₄
MDL Number:MFCD00027302
:110.2 g/mol
Boiling Pt:137 °C (760 mmHg)
Density:0.759 g/cm³
Flash Pt:18 °C
Storage Temperature:Ambient
UN:3295
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4270 to 1.4300 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:Approx. 200 ppm