2-(4-Nitrophenyl)ethanol 99%
:100-27-6
:
:C₈H₉NO₃
:MFCD00010202
:167.16 g/mol
:148…149 °C (2Torr)
:56…57 °C
:Ambient
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- NMR (1H-NMR):Consistent with structure
- Specific optical rotation:+166° to +175° (20°C, 589 nm) (1 % in ethanol)