Order Entry
Finland
ContactUsLinkComponent
Cerium(IV) sulphate 99%
Cerium(IV) sulphate 99%
Certificates
undefined
Cerium(IV) sulphate 99%
Certificates

Specification Test Results

  • Appearance:
    Yellow Powder and/or chunks
  • Titration Iodometric:
    39.6 to 44.7 % (Ce)
  • Refractive index:
    1.5500 to 1.5540 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information