Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Boiling Pt:56.5 °C
Density:0.789 Kg/l
UN:1090
ADR:,Ⅱ
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Solubility:Water
- Refractive index:20/D 1,3591
- Appearance:Liquid
- Min. assay (G.C.):99,5%
- Identity according to pharmacopoeias:Passes test
- Density 25/25:≤0,789
- Density 20/20:0,790 - 0,793
- Appearance of solution:Passes test
- Acidity:Passes test
- Alkalinity:Passes test
- Insoluble matter in H₂O:Passes test
- Non-volatile matter:0,004%
- Reducing substance to KMnO₄:Passes test
- Residual solvents (Ph. Eur./USP):Passes test
- Methanol:0,05%
- 2-Propanol:0,05%
- Benzene:0,0002%
- Other impurities:0,05%
- Ethanol (G.C.):500 ppm
- Water (H₂O):0,3%