
Acetic acid ≥75% for biopharmaceutical production
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,II
:GHS05
Related Information
Certificates
Search by Lot Number28579512.pdf spec_VWRC6519.2500 spec_VWRC6519.2500 spec_VWRC6519.9025 spec_VWRC6519.9200
Product specification
More
Acetic Acid ≥75% for Biopharmaceutical Production
Additional Product Details
sds_VWRC6519