Order Entry
Finland
Orders LinkContactUsLinkComponent
Nitrofurantoin, Reference Standards

Nitrofurantoin, Reference Standards

Supplier:   USP
Nitrofurantoin, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1464001.
  • 1021703.
  • Description:
  • Nitrofurantoin
  • Nitrofurantoin Related Compound A
  • Synonym(s):
  • N-(Aminocarbonyl)-N-[([5-nitro-2-furanyl]-methylene)-amino]-glycine
  • CAS No.:
  • 67-20-9
  • 63981-22-6
  • Formula:
  • C₈H₆N₄O₅
  • C₈H₈N₄O₆

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1464001.
    Nitrofurantoin
    Nitrofurantoin
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 419,00 €
    419,00 €
    1 * 500 mg
       419,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1464001.
    Description
    Nitrofurantoin
    Synonym(s)
    CAS No.
    67-20-9
    Formula
    C₈H₆N₄O₅
  • 1021703.
    Nitrofurantoin Related Compound A
    Nitrofurantoin Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 410,00 €
    1 410,00 €
    1 * 20 mg
       1 410,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1021703.
    Description
    Nitrofurantoin Related Compound A
    Synonym(s)
    N-(Aminocarbonyl)-N-[([5-nitro-2-furanyl]-methylene)-amino]-glycine
    CAS No.
    63981-22-6
    Formula
    C₈H₈N₄O₆