Graphene, single layer
Glass bottle, 1 g
Catalog # ACRO453150010
Supplier: Thermo Fisher Scientific
Graphene, single layer
Catalog # ACRO453150010
Supplier: Thermo Fisher Scientific
CAS Number: 1034343-98-0
Jump to:
Product Details & Documents
- CAS Number:1034343-98-0
- MDL Number:MFCD00144065
- Melting Pt:3650 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Black Solid
- Additional info:1-5 atomic layer graphene nanosheets
- Surface area:650 to 750 m²/g
- Oxygen (O):7 to 7.5 %
- Conductivity:500 to 700 S/m
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



