Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Nitromethane 99+% for analysis

Flammable
Irritant
Health
Nitromethane 99+% for analysis
Certificates
Nitromethane 99+% for analysis

3 Options Available Below

Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:100…102 °C (760 mmHg)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3805 to 1.3825 (20°C, 589 nm)
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.005 %
  • Cadmium (Cd):
    ≤0.1 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.