Order Entry
Finland
Orders LinkContactUsLinkComponent
Fenofibrate, Reference Standards

Fenofibrate, Reference Standards

Supplier:   USP
Fenofibrate, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1269447.
  • 1269607.
  • 1269618.
  • 1269629.
  • Description:
  • Fenofibrate
  • Fenofibrate Related Compound A
  • Fenofibrate Related Compound B
  • Fenofibrate Related Compound C
  • Synonym(s):
  • (4-Chlorophenyl)(4-hydroxyphenyl)methanone
  • 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
  • 1-methylethyl 2-[[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy]-2-methylpropanoate
  • CAS No.:
  • 49562-28-9
  • 42019-78-3
  • 42017-89-0
  • 217636-48-1
  • Formula:
  • C₂₀H₂₁ClO₄
  • C₁₃H₉ClO₂
  • C₁₇H₁₅ClO₄
  • C₂₄H₂₇ClO₆

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1269447.
    Fenofibrate
    Fenofibrate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 419,00 €
    419,00 €
    1 * 200 mg
       419,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269447.
    Description
    Fenofibrate
    Synonym(s)
    CAS No.
    49562-28-9
    Formula
    C₂₀H₂₁ClO₄
  • 1269607.
    Fenofibrate Related Compound A
    Fenofibrate Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 280,00 €
    1 280,00 €
    1 * 25 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269607.
    Description
    Fenofibrate Related Compound A
    Synonym(s)
    (4-Chlorophenyl)(4-hydroxyphenyl)methanone
    CAS No.
    42019-78-3
    Formula
    C₁₃H₉ClO₂
  • 1269618.
    Fenofibrate Related Compound B
    Fenofibrate Related Compound B
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 280,00 €
    1 280,00 €
    1 * 25 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269618.
    Description
    Fenofibrate Related Compound B
    Synonym(s)
    2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
    CAS No.
    42017-89-0
    Formula
    C₁₇H₁₅ClO₄
  • 1269629.
    Fenofibrate Related Compound C
    Fenofibrate Related Compound C
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 280,00 €
    1 280,00 €
    1 * 25 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1269629.
    Description
    Fenofibrate Related Compound C
    Synonym(s)
    1-methylethyl 2-[[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy]-2-methylpropanoate
    CAS No.
    217636-48-1
    Formula
    C₂₄H₂₇ClO₆
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.