Cyclohexane, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
UN:1145
ADR:3,II
Specification Test Results
- Assay (by GC) (corrected for water):min. 99.0 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 2 ppm
- Substances Darkened by H2SO4:passes test
- Water (by KF, coulometric):max. 100 ppm
- Single Impurity Peak (pg/ml):max. 10
- Single Impurity Peak (ng/ml):max. 10
Specification Test Results
Related Information
Certificates
Search by Lot NumberVWR J.T.Baker® High Purity solvents and reagents