Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, Oxybutynin impurity A, Ph.Eur., RS
Organic reference standard, Oxybutynin impurity A, Ph.Eur., RS
Catalog # EDQMO0288005
Supplier:  EDQM
CAS Number:  
Organic reference standard, Oxybutynin impurity A, Ph.Eur., RS
Catalog # EDQMO0288005
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    1199574-71-4
  • Description:
    Oxybutynin impurity A
  • Formula:
    C₂₂H₃₅NO₃
  • Storage temperature:
    Ambient
  • Description:
    Oxybutynin impurity A

Specifications

About this item

  • In Monograph 1354