Human plasma, Crohn's disease (CD)
Catalog # PB006C-1-CD
CAS Number:
Specifications
- Pk:2 mL
- Description:Plasma, Crohn's disease (CD), cryopreserved
Specifications
About this item
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of Crohn's disease (CD). Collected from IRB-consented donors diagnosed with CD.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.