Order Entry
Finland
ContactUsLinkComponent
Human plasma, chronic lymphocytic leukemia (CLL)
Human plasma, chronic lymphocytic leukemia (CLL)
Catalog # PB006C-1-CLL
CAS Number:  
Human plasma, chronic lymphocytic leukemia (CLL)
Catalog # PB006C-1-CLL
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Description:
    Plasma, chronic lymphocytic leukemia (CLL), cryopreserved

Specifications

About this item

Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of chronic lymphocytic leukemia (CLL). Collected from IRB consented chronic lymphocytic leukemia (CLL) disease donors.

No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.

Storage: Store at −80 °C.

Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.