Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Hydroxyzine Related Compounds, Reference Standard

Supplier:   USP
Hydroxyzine Related Compounds, Reference Standard
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1333003.
  • 1333058.
  • 1334006.
  • Description:
  • Hydroxyzine Hydrochloride
  • Hydroxyzine Related Compound A
  • Hydroxyzine Pamoate
  • Synonym(s):
  • 1-[(4-Chlorophenyl)phenylmethyl]piperazine
  • CAS No.:
  • 2192-20-3
  • 303-26-4
  • 10246-75-0
  • Formula:
  • C₂₁H₂₉Cl₃N₂O₂
  • C₁₇H₁₉ClN₂
  • C₄₄H₄₁ClN₂O₇

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1333003.
    Hydroxyzine Hydrochloride
    Hydroxyzine Hydrochloride
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 419,00 €
    419,00 €
    1 * 500 mg
       419,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1333003.
    Description
    Hydroxyzine Hydrochloride
    Synonym(s)
    CAS No.
    2192-20-3
    Formula
    C₂₁H₂₉Cl₃N₂O₂
  • 1333058.
    Hydroxyzine Related Compound A
    Hydroxyzine Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 300,00 €
    1 300,00 €
    1 * 25 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1333058.
    Description
    Hydroxyzine Related Compound A
    Synonym(s)
    1-[(4-Chlorophenyl)phenylmethyl]piperazine
    CAS No.
    303-26-4
    Formula
    C₁₇H₁₉ClN₂
  • 1334006.
    Hydroxyzine Pamoate
    Hydroxyzine Pamoate
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 381,00 €
    381,00 €
    1 * 500 mg
       381,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1334006.
    Description
    Hydroxyzine Pamoate
    Synonym(s)
    CAS No.
    10246-75-0
    Formula
    C₄₄H₄₁ClN₂O₇