Fulvestrant, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1286650.
- 1286661.
- Description:
- Fulvestrant
- Fulvestrant System Suitability Mixture
- CAS No.:
- 129453-61-8
- Formula:
- C₃₂H₄₇F₅O₃S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1286650.FulvestrantSpecifications:
More
Specifications
Cat. No.1286650.DescriptionFulvestrantCAS No.129453-61-8FormulaC₃₂H₄₇F₅O₃S - 1286661.Fulvestrant System Suitability MixtureFulvestrant System Suitability MixtureUSP reference standardVial30 mgLog in for availability1 * 30 mg1 410,00 €Specifications:
More
Specifications
Cat. No.1286661.DescriptionFulvestrant System Suitability MixtureCAS No.Formula



