B. subtilis recombinant NAD synthetase (from E. coli)
Catalog # PRSI90-367
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:B. subtilis
- Storage conditions:Stable for at least 6 months after receipt when stored at –80 °C.
- Enzyme name:NAD Synthetase
- Enzyme activity:~0.3 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1umol NAD per min.
- Purity:>98% (SDS-PAGE)
- Sequence:Full-length NAD synthetase
- Formulation:Liquid, In 50 mM TRIS-HCl, pH 8, containing 75mM sodium chloride, 5% glycerol and 5mM DTT.
Specifications
About this item
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways. Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It was shown to be induced by heat shock, salt stress, oxidative stress, glucose limitation and oxygen limitation and is involved in biological processes like stress response and sporulation resulting in formation of a cellular spore. Catalytic activity: ATP + deamido-NAD+ + NH3 = AMP + diphosphate + NAD+
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.