Human recombinant Sentrin-specific protease 2 (from E. coli)
50 µG
Catalog # PRSI92-222
Supplier: ProSci Inc.
Human recombinant Sentrin-specific protease 2 (from <i>E. coli</i>)
Catalog # PRSI92-222
Supplier: ProSci Inc.
CAS Number:
Jump to:
Product Details & Documents
SENP2 is an enzyme that belongs to the peptidase C48 family. SENP2 is a protease that catalyses two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SUMO1 is a small ubiquitin-like protein that can be covalently conjugated to other proteins. It has been implicated as a down-regulator of CTNNB1 levels and may therefore be a modulator of the Wnt pathway.
Fusion-Tag: N-6 His, SUMO tag
This recombinant protein can be used for biological assays. For research use only.
Documents
75791-038
Datasheet
Specifications
Specifications
- Pk:50 µG
- Cat. No.:PRSI92-222
- Enzyme type:Recombinant
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Formulation:Supplied as a 0.2 µm filtered solution of 50 mM HEPES, 5% glycerol pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Purity:>95% as determined by reducing SDS-PAGE
- Tag sequence:N-6 His, SUMO tag
- Sequence:Asp363-Leu589
- Tested applications:Biological assays
- Species:Human
- Source:E. coli
- Enzyme name:Sentrin-specific Protease 2
- Molecular weight:39.2 kD



