Order Entry
Finland
ContactUsLinkComponent
Rat recombinant FGF2 (from E. coli)
Rat recombinant FGF2 (from E. coli)
Catalog # PRSI92-617
Supplier:  ProSci Inc.
CAS Number:  
Rat recombinant FGF2 (from E. coli)
Catalog # PRSI92-617
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    HBGF-2|FGF2|FGF Basic|bFGF|FGF-2|Fibroblast Growth Factor 2|Basic Fibroblast Growth Factor|Heparin-Binding Growth Factor 2|FGFB
  • Protein/peptide name:
    FGF2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.2 kD
  • Sequence:
    Ala11-Ser154
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

FGF-basic is a members of the Fibroblast Growth Factors (FGFs) family. The family constitutes a large family of proteins involved in many aspects of development including cell proliferation, growth, and differentiation. They act on several cell types to regulate diverse physiologic functions including angiogenesis, cell growth, pattern formation, embryonic development, metabolic regulation, cell migration, neurotrophic effects, and tissue repair. FGF-basic is a non-glycosylated heparin binding growth factor that is expressed in the brain, pituitary, kidney, retina, bone, testis, adrenal gland liver, monocytes, epithelial cells and endothelial cells. FGF-basic signals through FGFR 1b, 1c, 2c, 3c and 4.

This recombinant protein can be used for biological assays. For research use only.