Order Entry
Finland
ContactUsLinkComponent
Human recombinant Activating Transcription Factor 1 (from E. coli)
Human recombinant Activating Transcription Factor 1 (from E. coli)
Catalog # PRSI92-277
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Activating Transcription Factor 1 (from E. coli)
Catalog # PRSI92-277
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Cyclic AMP-dependent transcription factor ATF-1|ATF1|Activating transcription factor 1|cAMP-dependent transcription factor ATF-1
  • Protein/peptide name:
    Activating Transcription Factor 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.3 kD
  • Sequence:
    Met1-Val271
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Cyclic AMP-dependent transcription factor ATF-1(ATF1) which contains 1 bZIP (basic-leucine zipper) domain and 1 KID (kinase-inducible) domain, belongs to the bZIP family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. ATF1 binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AC][AG]-3'), a sequence present in many viral and cellular promoters. It also binds to the Tax-responsive element (TRE) of HTLV-I. ATF1 mediates PKA-induced stimulation of CRE-reporter genes, represses the expression of FTH1 and other antioxidant detoxification genes, triggers cell proliferation and transformation. ATF1 is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and CDK3. Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.