Human recombinant Gamma-Synuclein (from E. coli)
PRSI92-120
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:BCSG1|SNCG|Persyn|SR|PRSN|Gamma-Synuclein|Synoretin|Breast Cancer-Specific Gene 1 Protein
- Protein/peptide name:Gamma-Synuclein
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:13.6 kD
- Sequence:Met1-Asp127
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Gamma-Synuclein (SNCG) is a member of the Synuclein protein family. Gamma-Synuclein is mostly expressed in the peripheral nervous system and retina. Gamma-Synuclein plays a role in neurofilament network integrity and may be involved in modulating axonal architecture during development and in the adult. In addition, it may also function in modulating the keratin network in skin. SNCG expression in breast tumours has been as a marker for tumour progression. SNCG is also believed to be involved in the pathogenesis of neurodegenerative diseases.
- This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.