Order Entry
Finland
ContactUsLinkComponent
 
Human Recombinant GSTZ1 (from E. coli)
  PRSI92-007
 :  
Human Recombinant GSTZ1 (from E. coli)
  PRSI92-007
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    GSTZ1|Maleylacetoacetate Isomerase|Glutathione S-Transferase Zeta 1|GSTZ1-1|MAAI
  • Protein/peptide name:
    GSTZ1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.18 kD
  • Sequence:
    Met1-Ala216
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM TrisHCl, 1mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Maleylacetoacetate Isomerase (MAAI) belongs to the Glutathione S-Transferase super-family. MAAI encodes multifunctional enzymes in the detoxification of electrophilic molecules by conjugation with glutathione, for example, mutagens, carcinogens and several therapeutic drugs. MAAI is a bifunctional protein with low glutathione peroxidase activity with T-butyl and cumene hydroperoxides. MAAI can catalyze the glutathione dependent oxygenation of dichloroacetic acid to glyoxylic acid. But it has minimal glutathione-conjugating activity with 7-chloro-4-nitrobenz-2-oxa-1, ethacrynic acid 3-diazole and maleylacetoacetate isomerase activity.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag