Human recombinant REG4 (from cells)
Catalog # PRSI91-903
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Gastrointestinal secretory protein|Regenerating islet-derived protein 4|Regenerating islet-derived protein IV|REG4|REG-like protein|GISP|RELP
- Protein/peptide name:REG4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:19.2 kD
- Sequence:Asp23-Pro158
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
REG4 is a secreted contains one C-type lectin domain, high expressed in the gastrointestinal tract, including jejunum, ileum, appendix, pancreas and small intestine. REG4 can be up-regulated by mucosal injury from active Crohn’s disease or ulcerative colitis. In the acid environment, REG4 can maintain carbohydrate recognition activity. REG4 may be involved in inflammatory and metaplastic response of the gastrointestinal epithelium.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag