Order Entry
Finland
ContactUsLinkComponent
Human recombinant Reg3 gamma (from cells)
Human recombinant Reg3 gamma (from cells)
Catalog # PRSI91-855
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Reg3 gamma (from cells)
Catalog # PRSI91-855
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Regenerating islet-derived protein III-gamma|Pancreatitis-associated protein 1B|REG3G|PAP-1B|REG-3-gamma|Regenerating islet-derived protein 3-gamma
  • Protein/peptide name:
    Reg3 gamma
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Glu27-Asp175
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Pancreatitis-associated protein IB, Regenerating islet-derived protein III-gamma, is a secreted protein which contains 1 C-type lectin domain. It is expressed almost exclusively in the pancreas and also expressed in testis, but not found in small intestine. This protein might be a stress protein involved in the control of bacterial proliferation.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag