Order Entry
Finland
ContactUsLinkComponent
Mouse recombinant CXCL12 (from E. coli)
Mouse recombinant CXCL12 (from E. coli)
  PRSI91-583
 :  
Mouse recombinant CXCL12 (from E. coli)
  PRSI91-583
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Gene ID:
    20315
  • Protein synonyms:
    TPAR1|Cxcl12|12-O-tetradecanoylphorbol 13-acetate repressed protein 1|Pre-B cell growth-stimulating factor|Stromal cell-derived factor 1|SDF-1 alpha|PBSF|C-X-C motif chemokine 12|SDF-1|Sdf1|Thymic lymphoma cell-stimulating factor|TLSF
  • Protein/peptide name:
    CXCL12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.1 kD
  • Sequence:
    Lys22-Lys89
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Mouse Cxcl12 is a secreted and highly conserved protein which belongs to the intercrine alpha (chemokine CxC) family.CXCL12 is widely expressed in various organs including brain, kidney, skeletal muscle, heart, liver, and lymphoid organs. Cxcl12 activates the C-X-C chemokine receptor CXCR4 to induce a rapid and transient rise in the level of intracellular calcium ions and chemotaxis. It also binds to atypical chemokine receptor ACKR3 which activates the beta-arrestin pathway and acts as a scavenger receptor for SDF-1. Cxcl12 has several critical functions during embryonic development such as B-cell lymphopoiesis, myelopoiesis in bone marrow and heart ventricular septum formation. Cxcl12 plays an important role in acting as a positive regulator of monocyte migration and a negative regulator of monocyte adhesion via the LYN kinase. It stimulates migration of monocytes and T-lymphocytes through its receptors, CXCR4 and ACKR3, and decreases monocyte adherence to surfaces coated with ICAM-1, a ligand for beta-2 integrins. SDF1A/CXCR4 signaling axis inhibits beta-2 integrin LFA-1 mediated adhesion of monocytes to ICAM-1 through LYN kinase. It also plays a protective role after myocardial infarction, induces down-regulation and internalization of ACKR3 expressed in various cells and stimulates the proliferation of bone marrow-derived b progenitor cells in the presence of IL-7 as well as growth of the stromal cell-dependent B-cell clone DW34 cells.

  • This recombinant protein can be used for biological assays. For research use only.