Human recombinant REG1A (from cells)
PRSI91-480
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:PSP|REG1A|REG-1-Alpha|Regenerating Islet-Derived Protein 1-Alpha|PTP|ICRF|Islet of Langerhans Regenerating Protein|Pancreatic Stone Protein|PSPS|Regenerating Protein I Alpha|Lithostathine-1-Alpha|Islet Cells Regeneration Factor|PSPS1|Pancreatic Thread Protein|REG
- Protein/peptide name:REG1A
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:17.31 kD
- Sequence:Gln23-Asn166
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Regenerating Islet-Derived Protein 1- alpha (REG1A) belongs to the Reg family of secreted proteins with a C-type lectic domain. REG1A is highly expressed levels in fetal and infant brains, much lower in adult brains. REG1A promotes the maintenance and growth of pancreatic islet cells and intestinal villi. In addition to, REG1A Might act as an inhibitor of spontaneous calcium carbonate precipitation and be associated with neuronal sprouting in brain, and with brain and pancreas regeneration.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag