Order Entry
Finland
ContactUsLinkComponent
Human recombinant ZBP1 (from E. coli)
Human recombinant ZBP1 (from E. coli)
Catalog # PRSI91-262
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ZBP1 (from E. coli)
Catalog # PRSI91-262
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Z-DNA-Binding Protein 1|DLM1|ZBP1|Tumor Stroma and Activated Macrophage Protein DLM|C20orf183
  • Protein/peptide name:
    ZBP1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Met1-Ser149
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Z-DNA Binding Protein 1 (ZBP1) is a protein with 2 DRADA repeats. ZBP1 is highly expressed in lymphatic tissues including lymph node, leukocytes, tonsil, bone marrow, and spleen. ZBP1 participates in the detection of viral and bacterial DNA from by the host's innate immune system. It plays a role in host defense against tumors and pathogens. ZBP1 Acts as a cytoplasmic DNA sensor which, when activated, induces the recruitment of TBK1 and IRF3 to its C-terminal region and activates the downstream interferon regulatory factor (IRF) and NF-kappa B transcription factors, leading to type-I interferon production. ZBP1-induced NF-kappaB activation probably involves the recruitment of the RHIM containing kinases RIPK1 and RIPK3.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag