Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Supelco® Drug and metabolite standards, ofloxacin, LoGiCal®
Supelco® Drug and metabolite standards, ofloxacin, LoGiCal®
PROOLGCFOR0228.00
:
LGC Standards PROMOCHEM
:
Supelco® Drug and metabolite standards, ofloxacin, LoGiCal®
PROOLGCFOR0228.00
:
LGC Standards PROMOCHEM
:
Pk:
10 mg
Standards:
Reference standard
CAS No.:
82419-36-1
Description:
Ofloxacin
Formula:
C₁₈H₂₀FN₃O₄
Description:
Ofloxacin