Anti-DLK1 Rat Monoclonal Antibody [clone: PF105B]
Catalog # PRSI36-059
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Delta-like 1 Homolog (Drosophila)
- Antigen symbol:DLK1
- Clonality:Monoclonal
- Clone:PF105B
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:13386
- Antigen synonyms:Fetal Antigen 1|Delta1|PREF1|Pref-1|FA1|ZOG|DLK|Protein Delta Homolog 1|DLK-1|Preadipocyte Factor 1|pG2
- Storage buffer:0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage temperature:Stable for at least 1 year after receipt when stored at -20˚C.
- Concentration:~1 mg/ml
- Shipping temperature:-20°C
- Immunogen:Recombinant mouse DLK1 (extracellular domain).
- Size:100 μg
- Pk:100 µG
Specifications
About this item
DLK1 (Protein delta homolog 1; Preadipocyte factor 1; Pref-1) is a regulator of adipocyte differentiation found in serum and urine. DLK1 might be a decoy ligand. Serum or plasma levels of soluble DLK1 affect negatively or positively adipogenesis and control mesenchymal cell fate. DLK1 is frequently upregulated in myelodysplastic syndrome (MDS) patients compared to non-leukemic individuals.
Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: PF105B
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse