Anti-ADRB2 Rabbit Polyclonal Antibody
PRSI80-019
- Antibody type:Primary
- Antigen name:Adrenoceptor Beta 2
- Antigen symbol:ADRB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:154
- Antigen synonyms:BETA2AR|ADRBR|Beta-2 adrenoreceptor|BAR|ADRB2R|Beta-2 adrenoceptor|B2AR
- Modification:phospho-Ser346
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:40 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:-20°C
- Immunogen:Adrenergic Receptor beta2 (phospho Ser346) antibody was raised against a peptide sequence around phosphorylation site of serine 346 (R-S-S (p)-L-K) derived from Human Adrenergic Receptor beta2.
- Purification:Purified by affinity-chromatography using epitope-specific peptide.
- Size:100 µg
- Pk:100 µG
Beta-adrenergic receptors mediate the catecholamine-induced activation of adenylate cyclase through the action of G proteins. The beta-2-adrenergic receptor binds epinephrine with an approximately 30-fold greater affinity than it does norepinephrine.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human