Anti-EIF2AK2 Rabbit Polyclonal Antibody
Product Details & Documents
Following activation by double-stranded RNA in the presence of ATP, the kinase becomes autophosphorylated and can catalyze the phosphorylation of the translation initiation factor EIF2S1, which leads to an inhibition of the initiation of protein synthesis. Double-stranded RNA is generated during the course of a viral infection.
Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10071-824.pdf
Datasheet
Specifications
- Size:100 µg
- Cat. No.:PRSI79-399
- Clonality:Polyclonal
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:double-stranded RNA-activated protein kinase|PRKR|Interferon-induced|EIF2AK1|eIF-2A protein kinase 2|Eukaryotic translation initiation factor 2-alpha kinase 2|PKR
- Immunogen:PKR (Ab-451) antibody was raised against a peptide sequence around aa.449~453 (K-G-T-L-R) derived from Human PKR.
- ImmunoFluorescence:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Concentration:1 mg/mL
- Antigen symbol:EIF2AK2
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



