Anti-SCN2B Rabbit Polyclonal Antibody
Catalog # PRSI51-062
Supplier: ProSci Inc.
Specifications
- Antigen name:Sodium Channel Voltage-gated Type II Beta
- Antigen symbol:SCN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SCNB2|ATFB14
- Immunogen:SCN2B antibody was raised against a peptide near the N-Terminus of SCN2B (Rat).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
PBS, 0.01% sodium azide, 50% glycerol.
Store SCN2B antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.
Type:
Antigen: SCN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat