Order Entry
Finland
ContactUsLinkComponent
 
Anti-TMEM123 Rabbit Polyclonal Antibody
  PRSI49-323
undefined
Anti-TMEM123 Rabbit Polyclonal Antibody
  PRSI49-323

 

  • Antigen name:
    Transmembrane Protein 123
  • Antigen symbol:
    TMEM123
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit,
    Monkey,
    Gibbon,
    Gorilla,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    PORIM_HUMAN.|Pro oncosis receptor inducing membrane injury|KCT3|Serine/threonine rich receptor|Transmembrane protein 123|Keratinocytes associated transmembrane protein 3|PORMIN|PORIMIN|TMEM123
  • Immunogen:
    TMEM123 antibody was raised against amino acids 173 - 188 of TMEM123 (Human).
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Porimin was identified by a monoclonal antibody developed by immunizing mice with apoptotic cells. One of these antibodies, designated anti-Porimin (for pro-oncosis receptor inducing membrane injury), was found to directly induce a unique type of cell death in Jurkat cells by binding to a 110- kDa cell surface receptor on Jurkat cells. Anti-Porimin-mediated cell death was preceded by cell aggregation, formation of plasma membrane pores, and the appearance of membrane blebs. More important, these cells show neither DNA fragmentation nor apoptotic bodies, but display lethal damage of the cell membrane.

TMEM123 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMEM123
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human, Monkey, Rabbit