Anti-RAD17 Rabbit Polyclonal Antibody
Catalog # PRSI47-172
Supplier: ProSci Inc.
Specifications
- Antigen name:RAD17 homolog
- Antigen symbol:RAD17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RAD17Sp|RAD17 homolog|RF C activator 1 homolog|Rad17 like protein|Rad24|HRAD17|Cell cycle checkpoint protein|R24L
- Immunogen:Peptide corresponding to aa 1233-1249 of the human BACH1 protein.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Affinity-purified
PBS containing 0.02% sodium azide.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RAD17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human