Order Entry
Finland
ContactUsLinkComponent
Anti-THRA Goat Polyclonal Antibody
Anti-THRA Goat Polyclonal Antibody
Catalog # PRSI46-487
Supplier:  ProSci Inc.
undefined
Anti-THRA Goat Polyclonal Antibody
Catalog # PRSI46-487
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Thyroid Hormone Receptor, Alpha
  • Antigen symbol:
    THRA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    triiodothyronine receptor|alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog|MGC43240|alpha|ERBA1|ERBA-related 7|AR7|THRA|thyroid hormone receptor|avian)|avian erythroblastic leukemia viral (v-erb-a) oncogene homolog|MGC000261|NR1A1|THRA2|EAR7|c-ERBA-1|ERB-T-1|THRA1|ERBA
  • Concentration:
    500 ug/mL
  • Immunogen:
    THRA antibody was raised against a 12 amino acid synthetic peptide near the internal region of THRA.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

THRA antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

THRA antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: THRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human