Order Entry
Finland
ContactUsLinkComponent
 
Anti-TFEC Goat Polyclonal Antibody
Catalog # PRSI46-483
Supplier:  ProSci Inc.
undefined
Anti-TFEC Goat Polyclonal Antibody
Catalog # PRSI46-483
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Transcription Factor EC
  • Antigen symbol:
    TFEC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    TFE-C|bHLHe34|TCFEC|hTFEC-L|TFEC-L|TFECL
  • Concentration:
    500 ug/mL
  • Immunogen:
    TFEC antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of TFEC.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

TFEC antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TFEC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TFEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human