Anti-SNTG1 Goat Polyclonal Antibody
Catalog # PRSI46-461
Supplier: ProSci Inc.
Specifications
- Antigen name:Syntrophin Gamma 1
- Antigen symbol:SNTG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:SYN4|G1SYN
- Concentration:500 ug/mL
- Immunogen:Syntrophin gamma 1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of Syntrophin gamma 1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Syntrophin gamma 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
SyntropH in gamma 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SNTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: