Anti-SUV39H2 Goat Polyclonal Antibody
Catalog # PRSI46-451
Supplier: ProSci Inc.
Specifications
- Antigen name:Suppressor of Variegation 3-9 homolog 2
- Antigen symbol:SUV39H2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- ChIP:Yes
- Form:Liquid
- Antigen synonyms:suppressor of variation 3-9 (Drosophila) homolog 2 (Drosophila) |suppressor of variegation 3-9 homolog 2|FLJ23414|SUV39H2|RP11-2K17.2|KMT1|KMT1B|uppressor of variegation 3-9 homolog 2 (Drosophila)
- Concentration:500 ug/mL
- Immunogen:SUV39H2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of SUV39H2.
- Tested applications:Chromatin
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SUV39H2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
SUV39H2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SUV39H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: