Order Entry
Finland
ContactUsLinkComponent
Anti-STX1A / STX1B Goat Polyclonal Antibody
Anti-STX1A / STX1B Goat Polyclonal Antibody
Catalog # PRSI46-448
Supplier:  ProSci Inc.
undefined
Anti-STX1A / STX1B Goat Polyclonal Antibody
Catalog # PRSI46-448
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Syntaxin 1A / Syntaxin 1B
  • Antigen symbol:
    STX1A / STX1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    C1Q and collagen domain containing|adipocyte|adipose most abundant gene transcript 1|syntaxin a/b|syntaxin 1B|syntaxin 1B2|HPC-1|STX1|STX1B2|FP17469|STX1B|HGNC:11433|p35-1|STX1A|syntaxin 1B1|STX1B1|adiponectin|syntaxin 1A
  • Concentration:
    500 ug/mL
  • Immunogen:
    STX1A antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of STX1A.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

STX1A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

STX1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STX1A / STX1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse