Anti-STK35 Goat Polyclonal Antibody
Catalog # PRSI46-445
Supplier: ProSci Inc.
Specifications
- Antigen name:Serine/Threonine Kinase 35
- Antigen symbol:STK35
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CLP-36 interacting kinase|Clik1
- Concentration:500 ug/mL
- Immunogen:STK35 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of STK35.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
STK35 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
STK35 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STK35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human